1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17600.06
also known as L17600-06
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
Catalog numberL17600.06
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS85275-45-2
IUPAC Nametert-butyl (3R)-3-hydroxypiperidine-1-carboxylate
Molecular FormulaC10H19NO3
InChI KeyUIJXHKXIOCDSEB-MRVPVSSYSA-N
SMILESCC(C)(C)OC(=O)N1CCC[C@@H](O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to yellow
Assay (GC)≥96.0%
Water Content (Karl Fischer Titration)≤2.5%
It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
RUO – Research Use Only