1-Boc-indole, 97%, Thermo Scientific Chemicals
1-Boc-indole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-indole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17601.03
also known as L17601-03
Price (USD)/ Each
47.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
47.70
Add to cart
1-Boc-indole, 97%, Thermo Scientific Chemicals
Catalog numberL17601.03
Price (USD)/ Each
47.70
-
Add to cart
Chemical Identifiers
CAS75400-67-8
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5390-1.5450 @ 20°C
Appearance (Color)Clear colorless to yellow to orange
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only