In Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
In Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.
Solubility
Soluble in water.
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Liang Zhang.; Tianhao Meng.; Jie Wu. Palladium-Catalyzed Suzuki-Miyaura Cross-Couplings of Aryl Tosylates with Potassium Aryltrifluoroborates.J. Org. Chem. 2007, 72 (24), 9346-9349 .
Ana Bellomoa.; David Gonzalez.; Hèlio A. Stefani. Synthesis of unnatural cyclitols via a combined enzymatic-palladium catalysis approach.J. Organomet. Chem. 2008, 693 (6),1136-1142 .