Thermo Scientific Chemicals

7-Fluoroindole, 97%, Thermo Scientific Chemicals

Catalog number: L17621.MD
250 mg, Each
Thermo Scientific Chemicals

7-Fluoroindole, 97%, Thermo Scientific Chemicals

Catalog number: L17621.MD
250 mg, Each
Quantity
Catalog number: L17621.MD
also known as L17621-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
387-44-0
Form
Crystals or powder or crystalline powder or fused solid or flakes
Melting Point (clear melt)
61.0-70.0?C
Appearance (Color)
White to cream to yellow to pale brown
Assay (GC)
≥96.0%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text