Thermo Scientific Chemicals

4-(Ethylthio)benzaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: L17624.03
1 g, Each
Thermo Scientific Chemicals

4-(Ethylthio)benzaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: L17624.03
1 g, Each
Quantity
Catalog number: L17624.03
also known as L17624-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84211-94-9
IUPAC Name
4-(ethylsulfanyl)benzaldehyde
Molecular Formula
C9H10OS
InChI Key
LPRKZLUVZOFDDT-UHFFFAOYSA-N
SMILES
CCSC1=CC=C(C=O)C=C1
Form
Liquid
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms
Refractive Index
1.6230-1.6270 @ 20?C

Description

It is an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text