Thermo Scientific Chemicals

5-(Boc-amino)-1-pentanol, 96%, Thermo Scientific Chemicals

Catalog number: L17649.03
1 g, Each
Thermo Scientific Chemicals

5-(Boc-amino)-1-pentanol, 96%, Thermo Scientific Chemicals

Catalog number: L17649.03
1 g, Each
Quantity
Catalog number: L17649.03
also known as L17649-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75178-90-4
Assay (GC)
≥95.0%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Viscous liquid
Refractive Index
1.4515-1.4595 @ 20?C

Description

5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

Solubility
Difficult to mix in water.

Notes
Moisture sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text