5-(Boc-amino)-1-pentanol, 96%, Thermo Scientific Chemicals
5-(Boc-amino)-1-pentanol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(Boc-amino)-1-pentanol, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17649.06
also known as L17649-06
Price (USD)/ Each
213.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
213.00
Add to cart
5-(Boc-amino)-1-pentanol, 96%, Thermo Scientific Chemicals
Catalog numberL17649.06
Price (USD)/ Each
213.00
-
Add to cart
Chemical Identifiers
CAS75178-90-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormViscous liquid
Refractive Index1.4515-1.4595 @ 20?C
5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Boc-amino)-1-pentanol may be used as non-limiting linkers.

Solubility
Difficult to mix in water.

Notes
Moisture sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
RUO – Research Use Only
Diane M. Yamamoto.; Donald A. Upson.; David K. Linn.; Victor J. Hruby. Synthesis of specific deuterium labeled tyrosine and phenylalanine derivatives and their use in the total synthesis of [8-arginine]vasopressin derivatives: the separation of diastereomeric [8-arginine]vasopressin derivatives by partition chromatography.J. Am. Chem. Soc. 1977, 99 (5),1564-1570 .