Thermo Scientific Chemicals

Potassium 4-fluorophenyltrifluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: L17655.06
5 g, Each
Thermo Scientific Chemicals

Potassium 4-fluorophenyltrifluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: L17655.06
5 g, Each
Quantity
Catalog number: L17655.06
also known as L17655-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
192863-35-7
IUPAC Name
potassium trifluoro(4-fluorophenyl)boranuide
Molecular Formula
C6H4BF4K
InChI Key
PXKNUQDMYBUYSZ-UHFFFAOYSA-N
SMILES
[K+].FC1=CC=C(C=C1)[B-](F)(F)F
Appearance (Color)
White
Form
Crystalline powder
Elemental Analysis
Carbon, C: 33.53-37.82% (Theory 35.68%)
Assay (unspecified)
≥94.0% to ≤106.0%, based on EA results
Elemental Analysis
Potassium, K: 18.19-20.52% (Theory 19.36%)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text