Thermo Scientific Chemicals

Ethyl benzimidate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L17684.09
10 g, Each
Thermo Scientific Chemicals

Ethyl benzimidate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L17684.09
10 g, Each
Quantity
Catalog number: L17684.09
also known as L17684-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5333-86-8
Appearance (Color)
White
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder

Description

Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text