Thermo Scientific Chemicals

Dimethyl naphthalene-2,6-dicarboxylate, 99+%, Thermo Scientific Chemicals

Catalog number: L17689.14
25 g, Each
Thermo Scientific Chemicals

Dimethyl naphthalene-2,6-dicarboxylate, 99+%, Thermo Scientific Chemicals

Catalog number: L17689.14
25 g, Each
Quantity
Catalog number: L17689.14
also known as L17689-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
840-65-3
IUPAC Name
2,6-dimethyl naphthalene-2,6-dicarboxylate
Molecular Formula
C14H12O4
InChI Key
GYUVMLBYMPKZAZ-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC2=CC=C(C=C2C=C1)C(=O)OC
Appearance (Color)
White to pale cream or pale yellow
Assay (chromatographic, GC or HPLC)
≥99.0%
Melting Point (clear melt)
188-198?C
Form
Powder or crystals or crystalline powder or flakes

Description

Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.

Solubility
Soluble in hot toluene (very faint turbidity). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text