2-Chloro-4-nitropyridine, 98%, Thermo Scientific Chemicals
2-Chloro-4-nitropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-4-nitropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17699.03
also known as L17699-03
Price (USD)/ Each
61.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
61.10
Add to cart
2-Chloro-4-nitropyridine, 98%, Thermo Scientific Chemicals
Catalog numberL17699.03
Price (USD)/ Each
61.10
-
Add to cart
Chemical Identifiers
CAS23056-36-2
IUPAC Name2-chloro-4-nitropyridine
Molecular FormulaC5H3ClN2O2
InChI KeyLIEPVGBDUYKPLC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Cl)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)49.0-55.0?C
Appearance (Color)Pale yellow to yellow
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only