5-Fluoro-2-iodoaniline, 97%, Thermo Scientific Chemicals
5-Fluoro-2-iodoaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-iodoaniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17723.06
also known as L17723-06
Price (USD)/ Each
214.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
214.00
Add to cart
5-Fluoro-2-iodoaniline, 97%, Thermo Scientific Chemicals
Catalog numberL17723.06
Price (USD)/ Each
214.00
-
Add to cart
Chemical Identifiers
CAS255724-71-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Assay (GC)≥96.0%
Melting Point (clear melt)38.0-48.0?C
FormCrystals or powder or crystalline powder or granules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only