D-Glucurono-6,3-lactone acetonide, 98+%, Thermo Scientific Chemicals
D-Glucurono-6,3-lactone acetonide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Glucurono-6,3-lactone acetonide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L17765.18
also known as L17765-18
Price (USD)/ Each
211.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
211.00
Add to cart
D-Glucurono-6,3-lactone acetonide, 98+%, Thermo Scientific Chemicals
Catalog numberL17765.18
Price (USD)/ Each
211.00
-
Add to cart
Chemical Identifiers
CAS20513-98-8
IUPAC Name9-hydroxy-4,4-dimethyl-3,5,7,11-tetraoxatricyclo[6.3.0.0²,⁶]undecan-10-one
Molecular FormulaC9H12O6
InChI KeyBDBGJSXZKMTMGP-UHFFFAOYNA-N
SMILESCC1(C)OC2OC3C(O)C(=O)OC3C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)> 98.0%
Optical Rotation+52.5? ? 2? (c=1, chloroform)
Solution Test10% in THF is clear and colourless
Formcrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only