3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.
Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
J. R. Beck.; J. A. Yahner. Synthesis of 2-cyanophenyl thiocyanates and related disulfides by nitro displacement. A novel synthesis of 3-chloro-1,2-benzisothiazole. J. Org. Chem. 1978, 43 (8), 1604-1606.
R. John Xavier.; E. Gobinathb. Density functional theory study on characterization of 3-chloro-1,2-benzisothiazole. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2012, 91 248-255.