7-Bromo-1-hydroxyisoquinoline, 97%, Thermo Scientific Chemicals
7-Bromo-1-hydroxyisoquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Bromo-1-hydroxyisoquinoline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17800.03
also known as L17800-03
Price (USD)/ Each
96.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
96.70
Add to cart
7-Bromo-1-hydroxyisoquinoline, 97%, Thermo Scientific Chemicals
Catalog numberL17800.03
Price (USD)/ Each
96.70
-
Add to cart
Chemical Identifiers
CAS223671-15-6
IUPAC Name7-bromo-1,2-dihydroisoquinolin-1-one
Molecular FormulaC9H6BrNO
InChI KeyDSOKREQUHLPVFR-UHFFFAOYSA-N
SMILESBrC1=CC=C2C=CNC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (HPLC)≥96.0%
Appearance (Color)Pale cream to yellow to brown
Melting Point (clear melt)245.5-251.5?C
FormPowder
7-Bromo-1-hydroxyisoquinoline acts as a pharmaceutical intermediate. It is also used to prepare 1-amino-4-bromoisoquinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1-hydroxyisoquinoline acts as a pharmaceutical intermediate. It is also used to prepare 1-amino-4-bromoisoquinoline.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ndubaku, C. O.; Crawford, T. D.; Chen, H.; Boggs, J. W.; Drobnick, J.; Harris, S. F.; Jesudason, R.; McNamara, E.; Nonomiya, J.; Sambrone, A.; Schmidt, S.; Smyczek, T.; Vitorino, P.; Wang, L.; Wu, P.; Yeung, S.; Chen, J.; Chen, K.; Ding, C. Z.; Wang, T.; Xu, Z.; Gould, S. E.; Murray, L. J.; Ye, W. Structure-Based Design of GNE-495, a Potent and Selective MAP4K4 Inhibitor with Efficacy in Retinal Angiogenesis. ACS Med. Chem. Lett. 2015, 6 (8), 913-918.
  2. Tikad, A.; Akssira, M.; Massip, S.; Léger, J. M.; Jarry, C.; Guillaumet, G.; Routier, S. Regiocontroled SNAr and Palladium Cross-Coupling Reactions of 2,4,7-Trichloropyrido[3,2-d]pyrimidine. Eur. J. Org. Chem. 2012, 2012 (24), 4523-4532.
  3. Price, D. A.; James, K.; Osborne, S.; Harbottle, G. W. Selective fluorination of 1-hydroxyisoquinolines using Selectfluor. Tetrahedron Lett. 2007, 48 (41), 7371-7373.