Thermo Scientific Chemicals

3-Iodobenzylamine, 97%, Thermo Scientific Chemicals

Catalog number: L17801.06
5 g, Each
Thermo Scientific Chemicals

3-Iodobenzylamine, 97%, Thermo Scientific Chemicals

Catalog number: L17801.06
5 g, Each
Quantity
Catalog number: L17801.06
also known as L17801-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
696-40-2
IUPAC Name
1-(3-iodophenyl)methanamine
Molecular Formula
C7H8IN
InChI Key
LQLOGZQVKUNBRX-UHFFFAOYSA-N
SMILES
NCC1=CC=CC(I)=C1
Appearance (Color)
Clear colorless to white to yellow
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Refractive Index
1.6355-1.6405 @ 20?C

Description

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Air sensitive and light sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text