Ethyl 4-acetylbutyrate, 97%, Thermo Scientific Chemicals
Ethyl 4-acetylbutyrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-acetylbutyrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L17848.06
also known as L17848-06
Price (USD)/ Each
42.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
42.00
Add to cart
Ethyl 4-acetylbutyrate, 97%, Thermo Scientific Chemicals
Catalog numberL17848.06
Price (USD)/ Each
42.00
-
Add to cart
Chemical Identifiers
CAS13984-57-1
IUPAC Nameethyl 5-oxohexanoate
Molecular FormulaC8H14O3
InChI KeyMGPSIDGTLFKDEY-UHFFFAOYSA-N
SMILESCCOC(=O)CCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to very pale yellow
FormLiquid
Assay (GC)>96.0%
Refractive Index1.4245-1.4295 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only