Thermo Scientific Chemicals

Pyridine-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: L17854.03
1 g, Each
Thermo Scientific Chemicals

Pyridine-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: L17854.03
1 g, Each
Quantity
Catalog number: L17854.03
also known as L17854-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
181219-01-2
IUPAC Name
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Formula
C11H16BNO2
InChI Key
NLTIETZTDSJANS-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=NC=C1
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder

Description

It is employed in the synthesis of new pyridazino[4,5-b]indol-4-ones and pyridazin-3(2H)-one analogs as DYRK1A inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of new pyridazino[4,5-b]indol-4-ones and pyridazin-3(2H)-one analogs as DYRK1A inhibitors.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text