Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane, Thermo Scientific Chemicals
Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L17891.06
also known as L17891-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS99337-56-1
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Refractive Index1.4420 - 1.4500 @20?C
Appearance (Color)Clear, colourless to pale yellow
Assay (Non-aqueous acid-base Titration)50.0 - 55.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only