Methyl 7-phenylheptanoate, 98%, Thermo Scientific Chemicals
Methyl 7-phenylheptanoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 7-phenylheptanoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17905.06
also known as L17905-06
Price (USD)/ Each
293.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
293.00
Add to cart
Methyl 7-phenylheptanoate, 98%, Thermo Scientific Chemicals
Catalog numberL17905.06
Price (USD)/ Each
293.00
-
Add to cart
Chemical Identifiers
CAS101689-18-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4920-1.4960 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only