Thermo Scientific Chemicals

5-Formylfuran-2-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: L17920.06
5 g, Each
Thermo Scientific Chemicals

5-Formylfuran-2-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: L17920.06
5 g, Each
Quantity
Catalog number: L17920.06
also known as L17920-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27329-70-0
IUPAC Name
(5-formylfuran-2-yl)boronic acid
Molecular Formula
C5H5BO4
InChI Key
JUWYQISLQJRRNT-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(O1)C=O
Form
Powder
Assay (HPLC)
≥94.0%
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Proton NMR
Conforms to structure
Appearance (Color)
Pale brown to brown

Description

As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text