Cyclohexyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals
Cyclohexyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L17952.06
also known as L17952-06
Price (USD)/ Each
64.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
64.30
Add to cart
Cyclohexyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals
Catalog numberL17952.06
Price (USD)/ Each
64.30
-
Add to cart
Chemical Identifiers
CAS953-91-3
IUPAC Namecyclohexyl 4-methylbenzene-1-sulfonate
Molecular FormulaC13H18O3S
InChI KeyOHHPZPDQZMUTCA-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
Purity (DSC)≥96.0%
Assay (HPLC)≥96.0%
Melting Point (DSC)43.0-46.0?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only