Thermo Scientific Chemicals

N,N'-Dibenzylethylenediamine diacetate, 99%, Thermo Scientific Chemicals

Catalog number: L17974.22
100 g, Each
Thermo Scientific Chemicals

N,N'-Dibenzylethylenediamine diacetate, 99%, Thermo Scientific Chemicals

Catalog number: L17974.22
100 g, Each
Quantity
Catalog number: L17974.22
also known as L17974-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-75-8
IUPAC Name
bis(acetic acid); benzyl[2-(benzylamino)ethyl]amine
Molecular Formula
C20H28N2O4
InChI Key
MTRNNCLQPVCDLF-UHFFFAOYSA-N
SMILES
CC(O)=O.CC(O)=O.C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1
Form
Powder
Melting Point (clear melt)
113.0-119.0°C
Appearance (Color)
White to pale cream
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text