2-Nitrobenzeneboronic acid, 96%, Thermo Scientific Chemicals
2-Nitrobenzeneboronic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitrobenzeneboronic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17988.06
also known as L17988-06
Price (USD)/ Each
297.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
297.00
Add to cart
2-Nitrobenzeneboronic acid, 96%, Thermo Scientific Chemicals
Catalog numberL17988.06
Price (USD)/ Each
297.00
-
Add to cart
Chemical Identifiers
CAS5570-19-4
IUPAC Name(2-nitrophenyl)boronic acid
Molecular FormulaC6H6BNO4
InChI KeySFUIGUOONHIVLG-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or cream to brown
Assay (HPLC)≥95.0%
Identification (FTIR)Conforms
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only