Thermo Scientific Chemicals

Methyltri-n-butylammonium chloride, Aliquat 175 (75% aq. soln.), Thermo Scientific Chemicals

Catalog number: L18019.22
100 g, Each
Thermo Scientific Chemicals

Methyltri-n-butylammonium chloride, Aliquat 175 (75% aq. soln.), Thermo Scientific Chemicals

Catalog number: L18019.22
100 g, Each
Quantity
Catalog number: L18019.22
also known as L18019-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56375-79-2
IUPAC Name
tributyl(methyl)azanium iodide
Molecular Formula
C13H30IN
InChI Key
DXJLCRNXYNRGRA-UHFFFAOYSA-M
SMILES
[I-].CCCC[N+](C)(CCCC)CCCC
Form
Liquid
Refractive Index
1.4550-1.4590 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (Titration ex Chloride)
>73.0 to <78.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text