Thermo Scientific Chemicals

Tris(pentafluorophenyl)borane, 97%, Thermo Scientific Chemicals

Catalog number: L18054.06
5 g, Each
Thermo Scientific Chemicals

Tris(pentafluorophenyl)borane, 97%, Thermo Scientific Chemicals

Catalog number: L18054.06
5 g, Each
Quantity
Catalog number: L18054.06
also known as L18054-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1109-15-5
IUPAC Name
tris(2,3,4,5,6-pentafluorophenyl)borane
Molecular Formula
C18BF15
InChI Key
OBAJXDYVZBHCGT-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Appearance (Color)
White to pale cream or pale grey
Assay from Supplier's CofA
≥96.0%
Form
Powder
Fluorine-19 NMR
Conforms to structure

Description

Tris(pentafluorophenyl)borane is used as an excellent activator component in homogeneous Ziegler-Natta chemistry. It is also used as a catalyst for reductions, alkylations, hydrometallation reactions and catalyzed aldol-type reactions. It is a useful Lewis acid and catalyzes hydrosilylation of aldehydes. Further, it is used in olefin polymerization catalysis. It serves as a reagent in the preparation of organometallic complexes. In addition to this, it is useful as polymerization catalysts and used to study the heterolytic cleavage of dihydrogen in associated with tri-tert-butylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(pentafluorophenyl)borane is used as an excellent activator component in homogeneous Ziegler-Natta chemistry. It is also used as a catalyst for reductions, alkylations, hydrometallation reactions and catalyzed aldol-type reactions. It is a useful Lewis acid and catalyzes hydrosilylation of aldehydes. Further, it is used in olefin polymerization catalysis. It serves as a reagent in the preparation of organometallic complexes. In addition to this, it is useful as polymerization catalysts and used to study the heterolytic cleavage of dihydrogen in associated with tri-tert-butylphosphine.

Solubility
Soluble in hexane, chloroform, dichloromethane, toluene, and polar solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and trimethylaluminum.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text