Thermo Scientific Chemicals

4-Cyclohexylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: L18076.06
5 g, Each
Thermo Scientific Chemicals

4-Cyclohexylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: L18076.06
5 g, Each
Quantity
Catalog number: L18076.06
also known as L18076-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
374538-04-2
IUPAC Name
(4-cyclohexylphenyl)boronic acid
Molecular Formula
C12H17BO2
InChI Key
KNQVRFYNQWNYPU-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(C=C1)C1CCCCC1
Appearance (Color)
White to grey
Assay (Aqueous acid-base Titration)
≥97.5%
Form
Crystals or powder or crystalline powder
Proton NMR
Conforms to structure
Assay (HPLC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text