Thermo Scientific Chemicals

3-(3-Bromophenyl)propionic acid, 97%, Thermo Scientific Chemicals

Catalog number: L18080.06
5 g, Each
Thermo Scientific Chemicals

3-(3-Bromophenyl)propionic acid, 97%, Thermo Scientific Chemicals

Catalog number: L18080.06
5 g, Each
Quantity
Catalog number: L18080.06
also known as L18080-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42287-90-1
IUPAC Name
3-(3-bromophenyl)propanoic acid
Molecular Formula
C9H9BrO2
InChI Key
DWKWMFSWLCIMKI-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=CC(Br)=CC=C1
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Powder
Melting Point (clear melt)
69.0-78.0?C
Appearance (Color)
White to orange
Assay (GC)
≥96.0%

Description

It is used in the synthesis and characterization of monoisomeric 1,8,15,22-substituted (A3B and A2B2) phthalocyanines and phthalocyanine-fullerene dyads.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and characterization of monoisomeric 1,8,15,22-substituted (A3B and A2B2) phthalocyanines and phthalocyanine-fullerene dyads.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text