Methyl 3-methylthiophene-2-carboxylate, 99%, Thermo Scientific Chemicals
Methyl 3-methylthiophene-2-carboxylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-methylthiophene-2-carboxylate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L18093.06
also known as L18093-06
Price (USD)/ Each
297.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
297.00
Add to cart
Methyl 3-methylthiophene-2-carboxylate, 99%, Thermo Scientific Chemicals
Catalog numberL18093.06
Price (USD)/ Each
297.00
-
Add to cart
Chemical Identifiers
CAS81452-54-2
IUPAC Namemethyl 3-methylthiophene-2-carboxylate
Molecular FormulaC7H8O2S
InChI KeyBRWROFVPMUPMJQ-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)C=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5405-1.5435 @ 20?C
FormLiquid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only