Thermo Scientific Chemicals

(S)-(-)-2-Benzylsuccinic acid 1-methyl ester, 98+%, ee 98+%, Thermo Scientific Chemicals

Catalog number: L18265.03
1 g, Each
Thermo Scientific Chemicals

(S)-(-)-2-Benzylsuccinic acid 1-methyl ester, 98+%, ee 98+%, Thermo Scientific Chemicals

Catalog number: L18265.03
1 g, Each
Quantity
Catalog number: L18265.03
also known as L18265-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
182247-45-6
IUPAC Name
(3S)-3-benzyl-4-methoxy-4-oxobutanoate
Molecular Formula
C12H13O4
InChI Key
BUNMUVFKMIOEQU-JTQLQIEISA-M
SMILES
COC(=O)[C@H](CC([O-])=O)CC1=CC=CC=C1
Appearance (Color)
Clear to turbid yellow
Optical Rotation
-27? ± 3.5? (c=1 in methanol)
Assay (GC)
≥98.0%
Form
Viscous liquid
Enantiomeric excess
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text