4-Methyl-3-nitrobenzamide, 98%, Thermo Scientific Chemicals
4-Methyl-3-nitrobenzamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-3-nitrobenzamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L18270.14
also known as L18270-14
Price (USD)/ Each
113.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
113.00
Add to cart
4-Methyl-3-nitrobenzamide, 98%, Thermo Scientific Chemicals
Catalog numberL18270.14
Price (USD)/ Each
113.00
-
Add to cart
Chemical Identifiers
CAS19013-11-7
IUPAC Name4-methyl-3-nitrobenzamide
Molecular FormulaC8H8N2O3
InChI KeyYEUGEQUFPMJGCD-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1[N+]([O-])=O)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to yellow
Assay (HPLC)≥97.5%
FormPowder or granules
Melting Point (clear melt)162.0-168.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only