Thermo Scientific Chemicals

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%, Thermo Scientific Chemicals

Catalog number: L18359.03
1 g, Each
Thermo Scientific Chemicals

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%, Thermo Scientific Chemicals

Catalog number: L18359.03
1 g, Each
Quantity
Catalog number: L18359.03
also known as L18359-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17342-08-4
IUPAC Name
(5S)-5-(hydroxymethyl)pyrrolidin-2-one
Molecular Formula
C5H9NO2
InChI Key
HOBJEFOCIRXQKH-BYPYZUCNSA-N
SMILES
OC[C@@H]1CCC(=O)N1
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤1.5%
Appearance (Color)
White to pale cream to pale brown
Assay (HPLC)
≥97.5%
Optical Rotation
30.5 ± 0.1? (C=2 in ethanol)

Description

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text