1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl, Thermo Scientific Chemicals
1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18426.03
also known as L18426-03
Price (USD)/ Each
87.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
87.90
Add to cart
1-Acetylpiperidine-4-carbonyl chloride, 97%, may contain up to ca 1M free HCl, Thermo Scientific Chemicals
Catalog numberL18426.03
Price (USD)/ Each
87.90
-
Add to cart
Chemical Identifiers
CAS59084-16-1
IUPAC Name1-acetylpiperidine-4-carbonyl chloride
Molecular FormulaC8H12ClNO2
InChI KeyOHCPVLJEAHBMEG-UHFFFAOYSA-N
SMILESCC(=O)N1CCC(CC1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to cream to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only