Thermo Scientific Chemicals

3-Dimethylaminobenzoyl chloride hydrochloride, tech. 90%, Thermo Scientific Chemicals

Catalog number: L18433.03
1 g, Each
Thermo Scientific Chemicals

3-Dimethylaminobenzoyl chloride hydrochloride, tech. 90%, Thermo Scientific Chemicals

Catalog number: L18433.03
1 g, Each
Quantity
Catalog number: L18433.03
also known as L18433-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
117500-61-5
IUPAC Name
3-(dimethylamino)benzoyl chloride hydrochloride
Molecular Formula
C9H11Cl2NO
InChI Key
NUDQPATXNFWFNK-UHFFFAOYSA-N
SMILES
Cl.CN(C)C1=CC=CC(=C1)C(Cl)=O
Appearance (Color)
Pale cream to cream to yellow to brown
Assay (Titration ex Chloride)
≥88.0%
Form
Powder or lumps

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text