Thermo Scientific Chemicals

Dimethylaminoacetyl chloride hydrochloride, tech. 85%, Thermo Scientific Chemicals

Catalog number: L18475.06
5 g, Each
Thermo Scientific Chemicals

Dimethylaminoacetyl chloride hydrochloride, tech. 85%, Thermo Scientific Chemicals

Catalog number: L18475.06
5 g, Each
Quantity
Catalog number: L18475.06
also known as L18475-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60853-81-8
IUPAC Name
2-(dimethylamino)acetyl chloride hydrochloride
Molecular Formula
C4H9Cl2NO
InChI Key
MVAXDKDOPWPFML-UHFFFAOYSA-N
SMILES
Cl.CN(C)CC(Cl)=O
Appearance (Color)
White to yellow or orange to brown
Form
Powder
Assay (Titration ex Chloride)
>83.0%

Description

Dimethylaminoacetyl chloride hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylaminoacetyl chloride hydrochloride is used as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store un
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text