Aluminum chloride, 98+%, extra pure, anhydrous powder
Aluminum chloride, 98+%, extra pure, anhydrous powder
Aluminum chloride, 98+%, extra pure, anhydrous powder
Aluminum chloride, 98+%, extra pure, anhydrous powder
Aluminum chloride, 98+%, extra pure, anhydrous powder
Thermo Scientific Chemicals

Aluminum chloride, 98+%, extra pure, anhydrous powder

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number L18489.36
also known as L18489-36
Price (USD)
103.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
103.00
Each
Add to cart
Chemical Identifiers
CAS7446-70-0
IUPAC Namealuminium(3+) trichloride
Molecular FormulaAlCl3
InChI KeyVSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to grey
Assay from Supplier's CofA≥98.0 to ≤102.0%
Impurity contentChromium Cr: ≤10 ppm (non-U.S. sourced material)
Impurity contentIron (Fe): ≤100 ppm (0.01%)
Impurity content(Free) Aluminium Al: ≤150 ppm (0.015%)
View more
Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

Solubility
Soluble in hydrochloric acid, ethanol, chloroform and carbon tetrachloride.Slightly soluble in benzene.

Notes
Aluminum chloride is hygroscopic. It must be packed in a tightly closed container and stored in a cool, dry, and well-ventilated area. It is water-reactive and corrosive in nature. It reacts vigorously with bases.
RUO – Research Use Only

General References:

  1. Lozhkina, O. A.; Panov, M. S.; Logunov, L. S.; Tumkin, I. I.; Gordeychuk, D. I.; Kochemirovsky, V. A. Aluminum Chloride Reveals the Catalytic Activity Towards Laser-Induced Deposition of Copper from Water-Based Solutions. Int. J. Electrochem. Sci. 2015, 10, 6084-6091
  2. Miyake, M.; Fujii, H.; Hirato, T. Electroplating of Al on Mg alloy in a dimethyl sulfone-aluminum chloride bath. Surf. Coat. Technol. 2015, 277, 160-164.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link