Ethyl trans-4-oxo-2-butenoate is an intermediate used to synthesize pharmaceutically active molecules. It is also used in the asymmetric catalytic alkynylation of acetaldehyde, producing attractive synthons with broad applications, such as (+)-Tetrahydropyrenophorol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl trans-4-oxo-2-butenoate is an intermediate used to synthesize pharmaceutically active molecules. It is also used in the asymmetric catalytic alkynylation of acetaldehyde, producing attractive synthons with broad applications, such as (+)-Tetrahydropyrenophorol.
Solubility
Slightly soluble in water(2.6 g/L at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Xiaoyu Wu.; Xiaoyang Dai.; Linlin Nie.; Huihui Fang.; Jie Chen.; Zhongjiao Ren.; Weiguo Cao.; Gang Zhao. Organocatalyzed enantioselective one-pot three-component access to indoloquinolizidines by a Michael addition-Pictet-Spengler sequence. Chemical Communications. 2010, 46 (16), 2733-2735.
- Ming He , Justin R. Struble , and Jeffrey W. Bode. Highly Enantioselective Azadiene Diels-Alder Reactions Catalyzed by Chiral N-Heterocyclic Carbenes. J. Am. Chem. Soc. 2006, 128 (26), 8418-8420.