Chemical Identifiers
CAS136849-72-4
IUPAC Nameethyl cyano({[(dimethylamino)(dimethyliminiumyl)methoxy]imino})formate; tetrafluoroboranuide
Molecular FormulaC10H17BF4N4O3
InChI KeyFPQVGDGSRVMNMR-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CCOC(=O)C(=NOC(N(C)C)=[N+](C)C)C#N
View more