4-Bromo-3-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals
4-Bromo-3-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number L18514.03
also known as L18514-03
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
4-Bromo-3-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals
Catalog numberL18514.03
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS374790-97-3
IUPAC Name(4-bromo-3-fluorophenyl)boronic acid
Molecular FormulaC6H5BBrFO2
InChI KeyACLQPRPXJMWADE-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Br)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥94.0%
Appearance (Color)White to pale brown
Assay (Aqueous acid-base Titration)≥94.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only