Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate, 97%, ee 96%, Thermo Scientific Chemicals
Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate, 97%, ee 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate, 97%, ee 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L18553.06
also known as L18553-06
Price (USD)/ Each
185.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
185.00
Add to cart
Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate, 97%, ee 96%, Thermo Scientific Chemicals
Catalog numberL18553.06
Price (USD)/ Each
185.00
-
Add to cart
Chemical Identifiers
CAS90866-33-4
IUPAC Nameethyl (3R)-4-chloro-3-hydroxybutanoate
Molecular FormulaC6H11ClO3
InChI KeyZAJNMXDBJKCCAT-RXMQYKEDSA-N
SMILESCCOC(=O)C[C@@H](O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4505-1.4555 @20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only