Thermo Scientific Chemicals

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals

Catalog number: L18554.03
1 g, Each
Thermo Scientific Chemicals

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 98%, Thermo Scientific Chemicals

Catalog number: L18554.03
1 g, Each
Quantity
Catalog number: L18554.03
also known as L18554-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86728-85-0
IUPAC Name
ethyl (3S)-4-chloro-3-hydroxybutanoate
Molecular Formula
C6H11ClO3
InChI Key
ZAJNMXDBJKCCAT-YFKPBYRVSA-N
SMILES
CCOC(=O)C[C@H](O)CCl
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4510-1.4550 @ 20?C
Form
Liquid
Optical Rotation
-14.0? ±1.0? (neat)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text