(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%, Thermo Scientific Chemicals
(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L18556.06
also known as L18556-06
Price (USD)/ Each
72.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
72.40
Add to cart
(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%, Thermo Scientific Chemicals
Catalog numberL18556.06
Price (USD)/ Each
72.40
-
Add to cart
Chemical Identifiers
CAS60827-45-4
IUPAC Name(2S)-3-chloropropane-1,2-diol
Molecular FormulaC3H7ClO2
InChI KeySSZWWUDQMAHNAQ-GSVOUGTGSA-N
SMILESOC[C@H](O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)>97.5%
Refractive Index1.4775-1.4825 @ 20?C
FormLiquid
Optical Rotation+1.2 ? 0.5?C (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only