Thermo Scientific Chemicals

(R)-(-)-Glycidyl butyrate, 98%, Thermo Scientific Chemicals

Catalog number: L18557.06
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-Glycidyl butyrate, 98%, Thermo Scientific Chemicals

Catalog number: L18557.06
5 g, Each
Quantity
Catalog number: L18557.06
also known as L18557-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
60456-26-0
IUPAC Name
[(2R)-oxiran-2-yl]methyl butanoate
Molecular Formula
C7H12O3
InChI Key
YLNSNVGRSIOCEU-ZCFIWIBFSA-N
SMILES
CCCC(=O)OC[C@H]1CO1
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4260-1.4300 @ 20?C
Optical Rotation
-32? ± 1.0? (neat)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text