3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L18567.14
also known as L18567-14
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
Add to cart
3,5-Dihydroxytoluene, 99%, Thermo Scientific Chemicals
Catalog numberL18567.14
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS504-15-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown or pale pink
FormCrystalline powder or powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point105-110°C
Used in production of orcein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only