Thermo Scientific Chemicals

2-Cyanoethyl diisopropylchlorophosphoramidite, 95%, Thermo Scientific Chemicals

Catalog number: L18575.06
5 g, Each
Thermo Scientific Chemicals

2-Cyanoethyl diisopropylchlorophosphoramidite, 95%, Thermo Scientific Chemicals

Catalog number: L18575.06
5 g, Each
Quantity
Catalog number: L18575.06
also known as L18575-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89992-70-1
IUPAC Name
3-({[bis(propan-2-yl)amino](chloro)phosphanyl}oxy)propanenitrile
Molecular Formula
C9H18ClN2OP
InChI Key
QWTBDIBOOIAZEF-UHFFFAOYNA-N
SMILES
CC(C)N(C(C)C)P(Cl)OCCC#N
Form
Liquid
Appearance (Color)
Clear to turbid or hazy colorless to yellow
Assay from Supplier's CofA
≥94.0%

Description

2-Cyanoethyl diisopropylchlorophosphoramidite is a useful intermediate in organic synthesis. It acts as a protecting reagent in DNA synthesis. It is involved in the selective monophosphorylation of carbohydrates and nucleosides. It plays an essential role in the conversion of protected ribonucleosides to phosphoramidites. It is also utilized as phosphitylating reagent for 3'-hydroxyl groups in the synthesis of oligodeoxyribonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanoethyl diisopropylchlorophosphoramidite is a useful intermediate in organic synthesis. It acts as a protecting reagent in DNA synthesis. It is involved in the selective monophosphorylation of carbohydrates and nucleosides. It plays an essential role in the conversion of protected ribonucleosides to phosphoramidites. It is also utilized as phosphitylating reagent for 3′-hydroxyl groups in the synthesis of oligodeoxyribonucleotides.

Solubility
Miscible with chloroform and dichloromethane.

Notes
Reacts violently with water. Incompatible with alcohols and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text