Thermo Scientific Chemicals

2-Bromobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: L18581.03
1 g, Each
Thermo Scientific Chemicals

2-Bromobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: L18581.03
1 g, Each
Quantity
Catalog number: L18581.03
also known as L18581-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
244205-40-1
IUPAC Name
(2-bromophenyl)boronic acid
Molecular Formula
C6H6BBrO2
InChI Key
PLVCYMZAEQRYHJ-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC=C1Br
Appearance (Color)
White to off-white
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥97.5%
Proton NMR
Conform to structure

Description

2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text