4-(Trifluoromethoxy)thiophenol, 95%, Thermo Scientific Chemicals
4-(Trifluoromethoxy)thiophenol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethoxy)thiophenol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18674.03
also known as L18674-03
Price (USD)/ Each
71.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
71.80
Add to cart
4-(Trifluoromethoxy)thiophenol, 95%, Thermo Scientific Chemicals
Catalog numberL18674.03
Price (USD)/ Each
71.80
-
Add to cart
Chemical Identifiers
CAS169685-29-4
IUPAC Name4-(trifluoromethoxy)benzene-1-thiol
Molecular FormulaC7H5F3OS
InChI KeyJHVNSRJPBXPZJU-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
Refractive Index1.4720-1.4780 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only