Thermo Scientific Chemicals

Bis(neopentyl glycolato)diboron, 97%, Thermo Scientific Chemicals

Catalog number: L18675.03
1 g, Each
Thermo Scientific Chemicals

Bis(neopentyl glycolato)diboron, 97%, Thermo Scientific Chemicals

Catalog number: L18675.03
1 g, Each
Quantity
Catalog number: L18675.03
also known as L18675-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
201733-56-4
IUPAC Name
2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Formula
C10H20B2O4
InChI Key
MDNDJMCSXOXBFZ-UHFFFAOYSA-N
SMILES
CC1(C)COB(OC1)B1OCC(C)(C)CO1
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text