Bis(neopentyl glycolato)diboron, 97%, Thermo Scientific Chemicals
Bis(neopentyl glycolato)diboron, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(neopentyl glycolato)diboron, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L18675.06
also known as L18675-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS201733-56-4
IUPAC Name2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular FormulaC10H20B2O4
InChI KeyMDNDJMCSXOXBFZ-UHFFFAOYSA-N
SMILESCC1(C)COB(OC1)B1OCC(C)(C)CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only