Thermo Scientific Chemicals

1-(2-Tetrahydrofuroyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog number: L18704.14
25 g, Each
Thermo Scientific Chemicals

1-(2-Tetrahydrofuroyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog number: L18704.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L18704.14
also known as L18704-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63074-07-7
IUPAC Name
4-[(2S)-oxolane-2-carbonyl]piperazin-1-ium
Molecular Formula
C9H17N2O2
InChI Key
UKESBLFBQANJHH-QMMMGPOBSA-O
SMILES
O=C([C@@H]1CCCO1)N1CC[NH2+]CC1
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.5175-1.5225 @ 20?C
Assay (GC)
≥96.0%
Form
Liquid or viscous liquid

Description

An intermediate of terazosin hydrochloride. It has also been used as a reactant for the preparation of pyrazol-3-propanoic acid derivatives as inhibitors of leukotriene biosynthesis in human neutrophils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An intermediate of terazosin hydrochloride. It has also been used as a reactant for the preparation of pyrazol-3-propanoic acid derivatives as inhibitors of leukotriene biosynthesis in human neutrophils.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text